Description
本研究采用强阴离子交换柱IonPac AS15色谱柱,建立了一种直接进样分析硝酸试剂中痕量阴离子的方法。使用淋洗液发生器在线产生KOH淋洗液,优化了色谱条件,本方法重现性好,测定了68%浓硝酸中的杂质离子,样品中氯离子和硫酸根的检出限分别达0.1,0.2 mg/L,取得了令人满意的结果。| Market: | Chemical |
| Keywords: | 离子色谱(IC);浓硝酸;阴离子 |
| Matrix: | 浓硝酸 |
For Research Use Only. Not for use in diagnostic procedures.