Description
本方法采用离子排斥和阴离子交换两种分离机理,建立的离子色谱柱切换方法实现了对高纯氟化盐干扰基体的在线消除,无需复杂的样品前处理。用于测定高纯氟化盐试剂中的痕量杂质无机阴离子,操作简单,精密度和回收率均较好,能满足实际氟化盐样品的分析要求。本方法为氟化盐高纯试剂的研发和生产监测提供了一种简便、稳定、灵敏的新途径,具有较高的实用价值和应用前景。| Market: | Chemical |
| Keywords: | 离子色谱;柱切换;氟化盐;高纯试剂;阴离子 |
| Matrix: | 高纯试剂 |
For Research Use Only. Not for use in diagnostic procedures.