Find methods for your needs
AN_C-GC-27化妆品中氯丁醇的检测 Be the first to rate this application
ancgc27化妆品中氯丁醇的检测
Description
本实验参考出入境检验检疫行业标准SN/T1780-2006 采用ECD 检测器检测化妆品的氯丁醇,方法快速灵敏,结果可靠,可以为化妆品中氯丁醇的检测提供参考。
Market: Chemical
Keywords: ECD;化妆品
Matrix: 化妆品
Uploaded on 10/14/2014.
For Research Use Only. Not for use in diagnostic procedures.