Description
为满足GB/T 3405 石油苯中对于噻吩含量的检测要求,采用ASTM D4735 需要配置PFPD 检测器,该检测器相对FPD价格昂贵,操作维护成本高。而采用Thermo Scientific TraceGC Ultra 配备了FPD 检测器后,测定精制苯中微量的噻吩,其检测结果完全满足GB/T 3405 石油苯中对于噻吩含量的要求,节省成本,操作简单,维护成本低,提高了实验室工作效率。| Market: | Chemical |
| Keywords: | 苯;噻吩;气相色谱 |
| Matrix: | 苯 |
For Research Use Only. Not for use in diagnostic procedures.