Description
采用ASE对土壤中的PAHs进行萃取,16种PAHs的加标回收率为86.7-116.2%,说明ASE适合萃取土壤中的PAHs。采用ASE萃取,一个样品前处理只需20 min,40 mL溶剂即可完成,具有简单、快速、高效和节省溶剂等特点。| Market: | Environmental |
| Keywords: | 加速溶剂萃取;气质联用;土壤;多环芳烃 |
| Matrix: | 土壤 |
For Research Use Only. Not for use in diagnostic procedures.