Description
离子色谱法测废水中的酸性红26,方法简便、稳定,线性范围内相关性好,准确度高,受其他因素干扰小。这类染料极性大,HPLC色谱柱很难保留,其结构也决定了质谱灵敏度很低,据文献报道LC-MS联用可检测限约为0.10 mg/L。而在上述色谱条件下,酸性红的检测限可到0.03 mg/L,离子色谱表现出一定的优势。Market: | Environmental |
Keywords: | 离子色谱;废水;酸性红26 |
Matrix: | 废水 |
For Research Use Only. Not for use in diagnostic procedures.