Description
本文采用在线固相萃取 - 超快速液相色谱法测定了水体中痕量甲萘威和呋喃丹,5分钟即可完成一次实际样品测定。样品无须经过繁琐的前处理过程,过滤后即可进样,样品的萃取和浓缩完全由液相色谱系统自动完成。与紫外检测器相比,串联质谱检测大大提高了检测灵敏度,适用于水体中超痕量甲萘威和呋喃丹的检测,样品需求量小,操作简单,回收率高| Market: | Environmental |
| Keywords: | 在线固相萃取;双三元;呋喃丹;甲萘威;水 |
| Matrix: | 水 |
For Research Use Only. Not for use in diagnostic procedures.