Description
采用一种新型双梯度液相色谱结合在线固相萃取技术建立了一种简便、快速、准确测定水中9种有机化合物的方法,本方法实现了样品在线富集及除杂,避免了繁琐的样品前处理过程,大大节约了样品量及时间,且重现性好,适用于水中微囊藻毒素-LR、甲萘威、莠去津、呋喃丹、百菌清、2,4,6-三氯酚、五氯酚、溴氰菊酯和苯并(a)芘等的测定。| Market: | Environmental |
| Keywords: | 双梯度高效液相色谱;微囊藻毒素;甲萘威;莠去津;呋喃丹;百菌清;三氯酚;五氯酚;溴氰菊酯;苯并芘 |
| Matrix: | 水 |
For Research Use Only. Not for use in diagnostic procedures.