Find methods for your needs
Improved analysis of budesonide and related substances using a Thermo Scientific Acclaim 300 C18 HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of budesonide and related impurities. The separation is performed on a Thermo Scientific Acclaim 300 C18 HPLC column, with UV detection at 240 nm.
Market: Pharma
Keywords: Acclaim 300 C18, Budesonide, Epimers, HPLC, Impurities, UV detection
Matrix: Budesonide inhaler (after 3 days)
Uploaded on 2/10/2010.
For Research Use Only. Not for use in diagnostic procedures.