Find methods for your needs
AN 004403: Screening of per- and polyfluoroalkyl substances (PFAS) in cosmetics: Utilizing a new combustion-ion chromatography system for total organic fluorine (TOF) analysis Be the first to rate this application
Description
A streamlined TOF workflow using the Cindion C-IC system was developed for PFAS screening in cosmetic products. The method has a TF detection limit of approximately 0.91 ppm, well below the level of 100 ppm often used as a screening threshold for high-fluorine samples, and the method is highly accurate and precise for both TF and TIF determination.
Market: Chemical
Keywords: Dionex Inuvion IC system, Dionex IonPac AS24 column, Cindion combustion/absorption module, AB 2771, lipstick, mascara, organofluorine compounds, foundation, makeup
Matrix: cosmetics
Author: Jingli Hu, Neil Rumachik
Affiliation: Thermo Fisher Scientific
Uploaded on 5/18/2026.
For Research Use Only. Not for use in diagnostic procedures.