Find methods for your needs
Utilizing TSQ mass spectrometry to quantitate 106 drugs of abuse in urine
Description
The Thermo Scientific™ Vanquish™ Horizon UHPLC coupled with the Thermo Scientific™ TSQ Quantis™ Plus mass spectrometer is used for the separation and detection of drugs of abuse in urine matrix. The SPE extraction uses Thermo Scientific™ SOLAμ™ SCX SPE plates for minimal sample and solvent volumes. Using a Thermo Scientific™ Accucore™ C18+ column with a total run time of 7 minutes, all isomers were separated including codeine / hydrocodone, and hydromorphone / morphine / norhydrocodone. The method includes 106 of the most tested drugs of abuse for easy adaption to customer labs.| Market: | Clinical Research |
| Keywords: | drugs of abuse, Toxicology, Urine, clinical, Forensics |
| Matrix: | Urine |
For Research Use Only. Not for use in diagnostic procedures.