Find methods for your needs
A ready-to-run workflow for quantitating 80 drugs of abuse in whole blood with the TSQ Certis Triple Quadrupole MS Be the first to rate this application
Description
The Thermo Scientific™ Vanquish™ Flex UHPLC coupled with the Thermo Scientific™ TSQ Certis™ mass spectrometer is used for the separation and detection of drugs of abuse in whole blood. The method features protein precipitation and filtration extraction protocol using ToT technology. Using a Thermo Scientific™ Accucore™ Biphenyl column with a total run time of 4.5 minutes, all isomers were separated, including methamphetamine / phentermine and hydromorphone / morphine / norcodeine. With >900 SRMs per second and sub 5 ms polarity switching, this method offers the speed and sensitivity required.
Market: Clinical Research
Keywords: drugs of abuse, Toxicology, clinical, Forensics, whole blood
Matrix: whole blood
Uploaded on 3/5/2026.
For Research Use Only. Not for use in diagnostic procedures.