Find methods for your needs
AB004183: Dionex CarboPac PA300-4um: A high-efficiency, high-resolution, all-purpose carbohydrate column
Description
This application brief demonstrates the performance of the Thermo Scientific Dionex CarboPac PA300-4um column for high-efficiency, high-resolution carbohydrate analysis using HPAE-PAD. The column effectively separates neutral and charged carbohydrates, including sugars, inulin, and alditols. Results show this product as an ideal first-pass column for carbohydrate method development, complementary to the CarboPac PA200.| Market: | Food and Beverage; BioPharma |
| Keywords: | Carbohydrates, Glycans, Monosaccharides, Sugars |
| Author: | Yukiko Kawahara, Neil Rumachik |
For Research Use Only. Not for use in diagnostic procedures.