Find methods for your needs
AN22032:Determination of four organotin compounds in water samples(联用测定水质样品中4种有机锡化合物) Be the first to rate this application
an22032determination four organotin compounds water samples(联用测定水质样品中4种有机锡化合物)
Description
In this application, a direct injection method for the determination of four organotin compounds in environmental water samples has been developed.(建立了直接进样测定环境水样中四种有机锡化合物的方法。) This method is using the Thermo Scientific U3000 HPLC and Thermo Scientific iCAP RQ ICP-MS.(采用U3000 HPLC 与iCAP RQ ICP-MS联用。) For the separation, a Thermo Scientific Acclaim 120 C18 column with isocratic elution was used.(使用Thermo Scientific Acclaim 120 C18色谱柱等度洗脱) An analysis can be completed within 12 minutes.(分析可在12分钟内完成。) This method is suitable for routine analysis in environmental industry.(适用于环境工业的常规分析。 )
Market: Environmental
Keywords: HPLC, ICPMS, organotin, 有机锡
Matrix: water(水)
Author: Ye Run(叶润);Li Xiaoli(李晓丽);Wang Fei(王飞)
Affiliation: Thermo Fisher Scientific
Uploaded on 5/16/2022.
For Research Use Only. Not for use in diagnostic procedures.