Find methods for your needs
AN21051 Determination of Diethylene Glycol in Oral Care Products by TG-WAXMS Column(TG-WAXMS应用于口腔护理产品中二甘醇的测定) Be the first to rate this application
diethylene glycol
Description
In this application, a method for Diethylene Glycol in Oral Care Products has been developed. This method is using the Thermo Scientific Trace 1310 GC-FID system. For the separation, a Thermo Scientific TG-WAXMS column with temperature gradient elution was used. An analysis can be completed within 18 minutes. This method is suitable for routine analysis in Consumer Goods industry and Testing Laboratory.
Market: Chemical
Keywords: diethylene glycol, Toothpaste, Oral Care Products, TG-WAXMS Column
Matrix: Mouthwash, Toothpaste
Author: Peihui Xu; Suzy Jin
Affiliation: Thermo Fisher Scientific
Uploaded on 5/16/2022.
For Research Use Only. Not for use in diagnostic procedures.