Find methods for your needs
Refine by Feature
More...
More...
More...
Name
More...
CAS Number
More...
Class
More...
Manufacturer
More...
Model
More...
Stationary Phase
More...
Dimensions
Length
Diameter/Tubing Size ID
Tubing Size OD
Particle Size/Film Thickness
Displaying 1-5 of 2510 results
Items per page:
Sort by
AN73883 Determination of per- and polyfluorinated alkyl substances (PFAS) in drinking water Using automated solid phase extraction and LC-MS/MS for U.S. EPA Method 533
Instrument Type: LCMSMS
Drinking water perfluorinated and polyfluorinated alkyl substances (PFAS) studies have typically targeted PFOS and PFBA and other PFAS. Here we discuss the development of an analytical method using an automated SPE system and LC-MS/MS for determination of twenty-five PFAS compounds following the guidelines provided by U.S. EPA Method 533. We have demonstrated that the automated SPE system provides reliable determination of PFAS in large-volume aqueous samples and saves time, solvent, and labor, while ensuring high reproducibility and productivity for analytical testing laboratories.
AN73883 Determination of PFAS in drinking water Using automated solid phase extraction and LC-MS/MS for U.S. EPA Method 533
Instrument Type: Automated Sample Prep
Drinking water perfluorinated and polyfluorinated alkyl substances (PFAS) studies have typically targeted PFOS and PFBA and other PFAS. Here we discuss the development of an analytical method using an automated SPE system and LC-MS/MS for determination of twenty-five PFAS compounds following the guidelines provided by U.S. EPA Method 533. We have demonstrated that the automated SPE system provides reliable determination of PFAS in large-volume aqueous samples and saves time, solvent, and labor, while ensuring high reproducibility and productivity for analytical testing laboratories.
AN002771 LC-MS/MS analysis of per- and polyfluoroalkyl substances (PFAS) in soil samples in accordance with EPA Method 1633: semi-automated solid phase extraction cleanup
Instrument Type: LCMS
PFAS workflows are continuously being developed to meet evolving testing requirements for per- and polyfluoroalkyl substances (PFAS) worldwide. This work describes that workflow by leveraging the Dionex AutoTrace 280 PFAS SPE instrument to automate the soil sample solid-phase extraction (SPE) cleanup stage that is described in EPA Method 1633.
AN002338: Quantitation of per- and polyfluoroalkyl substances (PFAS) in aqueous samples by LC-MS/MS following EPA Draft Method 1633
Instrument Type: LCMS
PFAS are per- and polyfluoroalkyl substances. The chemical nature of the C-F bonds makes these compounds extremely stable. Hence, PFAS have been given the term “forever compounds”. This application note will present data for measuring 40 PFAS in fortified water samples following the third draft of EPA Method 1633. An MDL study was conducted in reagent water to demonstrate that equivalent or better performance can be attained using the Thermo Scientific™ Vanquish™ Flex Binary UHPLC system and Thermo Scientific™ TSQ™ Quantis Plus mass spectrometer.
Chiral analysis of Vilanterol Trifenatate using a Hypersil Chiral IGT column
Instrument Type: UHPLC
The Thermo Scientific UltiMate 3000 RS HPLC System is applied for the chiral purity of (S)-Isomer in Vilanterol Trifenatate. The analysis was performed on a Thermo Scientific Hypersil Chiral IGT HPLC column using an in-house method. The result obtained for resolution between S-isomer and Vilanterol Trifenatate is 4.12.The results obtained for system suitability were 2.37 for the S-isomer and Vilanterol Trifenatate were 1.0 and 1.02 respectively.