Find methods for your needs
Untargeted analysis with GC-Orbitrap: a powerful tool for the authentication of spices and herbs Be the first to rate this application
untargeted analysis with gcorbitrap a powerful tool for authentication spices herbs
Description
In this study GC-Orbitrap technology coupled with solid-phase micro-extraction (SPME) with Arrow technology was used to assess the volatile profile of intentionally adulterated and native oregano samples.
Market: Food and Beverage
Keywords: GC, Adulteration, Orbitrap, Gas Chromatography, EI, electron ionization, ExtractaBrite, HRMS, fraud, full-scan, herbs, high resolution mass spectrometry, Oregano, PCI, positive chemical ionization, spices, unknowns, volatile organic compounds (VOCs)
Matrix: Oregano
Author: Giulia Riccardino1, Dominic Roberts1, Cristian Cojocariu1, and Michele Suman2
Affiliation: 1Thermo Fisher Scientific, Runcorn, UK; 2Barilla SpA – Advanced Research Laboratories, Parma, IT
Uploaded on 3/3/2021.
For Research Use Only. Not for use in diagnostic procedures.