Find methods for your needs
Dionex IonPac AS32-Fast-4µm 2mm Column Performance Test using its QAR Method Be the first to rate this application
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report (QAR). This record provides an eWorkflow for executing the QAR method on ICS-5000+ system. Other ICS systems can be used for this application.
Market: Environmental
Keywords: ICS-5000+, ICS-6000, ADRS 600 Suppressor, IonPac AS32-Fast-4µm column, QAR condition
Matrix: Water
Author: Jingli Hu and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 7/26/2019.
For Research Use Only. Not for use in diagnostic procedures.