Find methods for your needs
Thermo Scientific Dionex IonPac AS29-Fast-4µm Column Performance Test Using its QAR Method Be the first to rate this application
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report. At least three injections should be made. This record provides an eWorkflow for executing the QAR method on an ICS-5000+ system. An ICS-6000 can be used for this application.
Market: Environmental
Keywords: Ion Chromatography (IC), fluoride, Nitrate, Nitrite, Chloride, phosphate, Sulfate, Suppressed Conductivity, wastewater, Drinking water, Bromide, IonPac AS29-Fast-4µm column
Matrix: Drinking water, Wastewater
Author: Hua Yang; Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 7/11/2019.