Find methods for your needs
Rapid qualitative and quantitative analysis of residual solvents in food packaging by static headspace coupled to GC-FID/MS Be the first to rate this application
rapid qualitative quantitative analysis residual solvents food packaging by static headspace coupled gcfidms
Description
This application note demonstrates the qualitative and quantitative performance of the Thermo Scientific™ TriPlus™ 500 Gas Chromatography Headspace Autosampler coupled to a dual-detector GC-FID/MS for the determination of residual solvents in food packaging according to the European Standard EN 13628-1 method1 and to highlight a highly efficient workflow through extended automation from sampling to data reporting.
Market: Food and Beverage
Keywords: FID, MHE, Residual solvents, MS, Food Safety, Flame ionization detector, ISQ 7000, HS-GC, valve and loop, flexible food packaging, headspace-gas chromatography, multiple headspace extraction, mass spectrometer detector, single quadrupole GC-MS, TriPlus 500 HS
Matrix: Flexible food packaging
Author: Giulia Riccardino and Cristian Cojocariu
Affiliation: Thermo Fisher Scientific, Runcorn, UK
Uploaded on 5/13/2019.
For Research Use Only. Not for use in diagnostic procedures.