Find methods for your needs
Tandem NanoLC-MS: Maximum MS Utilization for Deep Dive Proteomic Analysis Be the first to rate this application
tandemnanolcms
Description
Here we show that with a modest hardware extension to the UltiMate 3000 RSLCnano platform, namely the addition of a second nano-flow high-pressure gradient pump and 1/32” nano post column switching valve plus the associated additional fluidics, columns and column switching valve, all of which are commercially available, a tandem nanoLC stack can be created which enables up to twice the throughput capacity compared to a single LC-MS system.
Market: Proteomics
Keywords: Proteomics, continuous MS utilization, nanoLC-MS, Tandem nano-flow LC
Matrix: cell lysate, HeLa protein digest
Author: Christopher Pynn, Alexander Boychenko, Wim Decrop, Mike Baynham, Peter Jehle, Martin Ruehl
Affiliation: Thermo Fisher Scientific
Uploaded on 11/21/2018.
For Research Use Only. Not for use in diagnostic procedures.