Find methods for your needs
Fast, ultra-sensitive analysis of PBDEs in food using advanced GC-MS/MS technology Be the first to rate this application
fast analysis 27 pbdes food by gcmsms
Description
The aim of the study was to assess the quantitative performance of the Thermo Scientific™ TSQ™ 9000 triple quadrupole GC-MS/MS system equipped with the advanced electron ionization (AEI) source for the analysis of polybrominated diphenyl ethers (PBDEs) in food at low concentrations in under 11 minutes on the TG-PBDE column.
Market: Food and Beverage
Keywords: Chromeleon, sensitivity, Food Safety, trace analysis, isotope dilution, Polybrominated Diphenyl Ethers (PBDEs), Gas Chromatography, triple quadrupole mass spectrometry, TSQ 9000, Advanced Electron Ionization (AEI), Routine, Selective Reaction Monitoring (SRM)
Matrix: Cows Liver, Fish Oil, Milk, Reindeer, Salmon, Tallow
Author: Aaron Lamb¹, Sean Panton², Cristian Cojocariu¹
Affiliation: ¹Thermo Fisher Scientific, Runcorn, UK, ²Fera Science, Ltd., York, UK
Uploaded on 10/15/2018.
For Research Use Only. Not for use in diagnostic procedures.