Find methods for your needs
Overcoming analytical challenges for polybrominated diphenyl ethers (PBDEs) analysis in environmental samples using gas chromatography – Orbitrap mass spectrometry Be the first to rate this application
pbdes using orbitrap mass spectrometry
Description
This work demonstrates the applicability of high-resolution, accurate-mass GC-Orbitrap technology for the targeted analysis of 27 PBDE congeners in environmental samples with variable complexity using a sensitive, fast, robust method. This approach takes into account the selectivity, sensitivity, linearity, reproducibility of the results, method robustness, and analysis time.
Market: Environmental
Keywords: PBDE, Polybrominated diphenyl ethers, environmental, Quantification, Accurate mass, GC Orbitrap, targeted SIM, high-resolution GC-MS
Matrix: Air, environmental, filter dust, Sediment, Sludge
Author: Jane Cooper(1) Jordi Parera(2) Esteban Ebad (2) Richard Law(1) and Cristian Cojocariu(1)
Affiliation: (1) Thermo Fisher Scientific, Runcorn, UK (2) Lab of Dioxins, Dept. of Environmental Chemistry, IDAEA, Spanish National Research Council (CSIC)
Uploaded on 10/8/2018.
For Research Use Only. Not for use in diagnostic procedures.