Rate
Ask the Expert
Suggest an Application
Add to Favorites
Create
Parts List
Parts List
Tell a
Friend
Friend
Print
PDF
Find methods for your needs
AN72751: Determination of Anionic Impurities in Sulfuric Acid using Ion Chromatography (Method 2)
Description
This study demonstrated that anionic impurities such as fluoride, chloride, nitrite, bromide, and nitrate can be determined accurately in sulfuric acid using Method 1. Method 1 uses a Dionex IonPac AS19 HC-4 µm column with hydroxide eluent, which is ideal for separating a wide range of inorganic anions with high sensitivity. The anionic impurity phosphate can be determined accurately in sulfuric acid using Method 2. Method 2 uses a Dionex IonPac AS23-4μm column with carbonate eluent, which is ideal for phosphate because it elutes before sulfate. This record is Method 2.Market: | Chemical; Electronics & Semiconductors |
Keywords: | Dionex IonPac AS23-4μm column, Dionex ICS 5000+ HPIC system, Dionex ICS 6000 HPIC system, suppressed conductivity detector |
Matrix: | Water |
Author: | Jingli Hu and Jeffrey Rohrer |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.