Find methods for your needs
Routine determination of ultratrace elements in semiconductor grade nitric acid by the Thermo Scientific iCAP RQ ICP-MS Be the first to rate this application
Description
To determine ultratrace metal concentrations in semiconductor grade nitric acid (HNO3). To demonstrate the use of cold plasma (CP) and kinetic energy discrimination (KED) to reduce background equivalent concentrations (BEC) and improve detection limits (LoD). Demonstrate the use of the Thermo Scientific™ iCAP™ RQ ICP-MS to perform reproducible ultratrace ng·L⁻¹ (ppt) measurements of semiconductor relevant elements in nitric acid.
Market: Electronics & Semiconductors
Keywords: ICP-MS, Cold plasma, iCAP RQ, nitic acid, semiconductor analysis
Matrix: HNO3, Nitric Acid
Author: Tomoko Vincent[1], Jones Hsu [2], Vicki Wu [3], Julian Wills [1]
Affiliation: [1] Thermo Fisher Scientific, Bremen, Germany; [2] BASF, Taipei, Taiwan; [3] Joy Allied Technology, Taipei, Taiwan
Uploaded on 2/16/2018.
For Research Use Only. Not for use in diagnostic procedures.