Find methods for your needs
Fipronil GC-MS/MS Be the first to rate this application
analysis fipronil eggsusing gcmsms
Description
In this study, the results obtained for the analysis of fipronil residue in egg using a Thermo Scientific triple quadrupole GC-MS system are reported. System performance was tested using full scan acquisition and simple instrumental setup. The experiments performed focused on assessing the sensitivity, linearity, selectivity and analytical precision for the analysis of fipronil and its metabolites in eggs.
Market: Food and Beverage
Keywords: sensitivity, Food Safety, Selected Reaction Monitoring, eggs, fipronil, fipronil desulfinyl, fipronil sulfone, triple quadrupole gas chromatography, TSQ 8000 Evo
Matrix: eggs
Author: Cristian Cojocariu1, Joachim Gummersbach2 and Susanne Sales2
Affiliation: 1Thermo Fisher Scientific, Runcorn, UK; 2Thermo Fisher Scientific, Dreieich, Germany
Uploaded on 9/8/2017.
For Research Use Only. Not for use in diagnostic procedures.