Find methods for your needs
A novel high resolution accurate mass Orbitrap-based GC-MS for routine analysis of Short Chained Chlorinated Paraffins Be the first to rate this application
orbitrap gcms analysis sccps
Description
Short Chained Chlorinated Paraffins (SCCPs) are emerging contaminants that, once released, will remain in the environment for long periods of time and therefore can bioaccumulate in living organisms. SCCPs are intentionally manufactured and used as lubricants and coolants in the metal processing industry or as plasticizers and flame retardants in plastic products. Chronic exposure to SCCPs is believed to have harmful and irreversible effects for humans and the environment [1]. As a consequence, SCCPs are listed in the Stockholm convention as chemicals with potential adverse effects and their p
Market: Environmental
Keywords: High resolution, sensitivity, Orbitrap Technology, Gas Chromatography, Persistent Organic Pollutants (POPs), emerging contaminants, Exactive GC, hort Chained Chlorinated Paraffins (SCCPs), mass accuracy, nvironment
Author: Cristian Cojocariu and Paul Silcock
Affiliation: Thermo Fisher Scientific, Runcorn, UK
Uploaded on 8/17/2017.
For Research Use Only. Not for use in diagnostic procedures.