Find methods for your needs
AN72349: Determination of Chlorine, Bromine, and Sulfur in Polyethylene Materials using Combustion Ion Chromatography Be the first to rate this application
Description
Plastic samples can be very difficult to analyze as sample preparation is required to extract analytes or remove interfering matrices, and these sample preparation techniques can be labor intensive. In this application note, we demonstrate that a CIC system can be used for automated qualitative and quantitative analysis of halogens and sulfur in plastic samples. Four plastic samples were analyzed for their chlorine (Cl), bromine (Br), and sulfur (S) content. Method accuracy was evaluated using a polymer certified reference material (ERM EC680k) for Cl,Br, and S.
Market: Environmental; Chemical
Keywords: IonPac AS11-HC-4μm column, Polymer, CIC, Sulfur, Chlorine, Bromine, Integrion HPIC, Combustion ion chromatography, plastic samples, Low density polyethylene, ERM EC680K, TN72211, AN 72349
Matrix: Plastic
Author: Manali Aggrawal and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 5/19/2017.
For Research Use Only. Not for use in diagnostic procedures.