Find methods for your needs
PN70733: Automated Solid-Phase Extraction of Phthalates for Drinking Water Samples Be the first to rate this application
pn70733 automated solidphase extraction phthalates for drinking water samples
Description
Phthalates are widely used in a large variety of consumer products to provide flexibility and durability to plastics such as polyvinyl chloride (PVC) including medical devices, children’s toys, personal care products, and food packaging. When used in PVC production phthalates tend to evaporate into the air and leach into water and soil over time. Humans and other living organisms can be exposed to these toxic phthalates and the U.S.EPA has written Method 506 for monitoring of these compounds. This application note provides a method for SPE and GC-ECD to determine phthalates in drinking water.
Market: Environmental
Keywords: Solid phase extraction, SPE, automted SPE, Benzylbutylphthalate (BBP), Bis(2-ethylhexyl) phthalate (DEHP), Dibutyl phthalate (DBP), Diisodecyl phthalate (DIDP), Diisononyl phthalate (DINP), Di-n-octyl phthalate (DNOP)
Matrix: Drinking water
Author: Alex Zhang, Pranathi Perati, Linda Lopez
Affiliation: Thermo Fisher Scientific
Status: Proof of Concept
Uploaded on 8/9/2016.
For Research Use Only. Not for use in diagnostic procedures.