Find methods for your needs
AN66: Determination of Neomycin B and Impurities Using HPAE-IPAD Be the first to rate this application
determination neomycin b
Description
In this application note, we combine the CarboPac PA1, an eluent generator with CR-ATC, and disposable Au working electrodes to demonstrate an improved HPAE-PAD technology for Neomycin B purity analysis and its determination in Neosporin topical ointment, a complex over-the-counter pharmaceutical formulation.
Market: BioPharma
Keywords: aminoglycoside, carbohydrate, CarboPac PA1, Electrochemical detection, Eluent Generation, nebramine, Pharmaceutical, neamine, ointment, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 66
Matrix: Ointment
Author: Valoran Hanko and Jeff Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 4/1/2015.
For Research Use Only. Not for use in diagnostic procedures.