Find methods for your needs
AN61: Determination of Tobramycin and Impurities Using HPAE-PAD Be the first to rate this application
determination tobramycin
Description
In this application note, we combine the CarboPac PA1, eluent generator with CR-ATC, and disposable Au working electrodes to demonstrate an improved HPAE-PAD technology for tobramycin purity analysis.
Market: Pharma
Keywords: aminoglycoside, aminoglycoside antibiotic, Antibiotic, carbohydrate, CarboPac PA1, Drug Substance, kanamycin, kanamycin B, nebramine, neamine, PAD, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 61
Matrix: Drug
Author: Valoran Hanko and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 2/17/2015.
For Research Use Only. Not for use in diagnostic procedures.