Find methods for your needs
AN270: Determination of Hydroxymethylfurfural (HMF) in Honey and Biomass Be the first to rate this application
determination hmf
Description
This work describes a high-performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD)-based method for the determination of HMF in samples ranging from food to treated biomass. A Dionex ICS-3000 system with a CarboPac PA1 column, electrolytically generated hydroxide eluent, and electrochemical detection with disposable Au-on-polytetrafluoroethylene working electrodes are used. The Dionex CarboPac PA1 is a high-capacity, rugged column suitable for determining mono- and disaccharides, and has high resolution for HMF in a wide variety of matrices.
Market: Food and Beverage
Keywords: 5-hydroxymethylfurfural, biofuel, biofuels, Carbohydrates, CarboPac PA1, corn stover, disposable electrodes, EG, Eluent Generation, food spoilage, HMF, hydroxymethylfurfural, syrup, Honey, PAD, HPAE-PAD, TN 20, TN 70671, TN20, TN70671, AN 270
Matrix: Biomass, Corn stover, Honey
Author: Lipika Basumallick and Jeff Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 2/3/2015.
For Research Use Only. Not for use in diagnostic procedures.