Description
采用赛默飞世尔新型的ISQ单四级杆气质联用仪检测农药残留,准确定量目标物,RSD值小,灵敏度高,无杂质峰干扰。ISQ无需泄真空就能够快速在EI、CI之间切换,使分析农残类化合物变得更简单。| Market: | Other |
| Keywords: | 气质联用;负化学源;茶叶;有机氯;拟除虫菊酯 |
| Matrix: | 茶叶 |
For Research Use Only. Not for use in diagnostic procedures.