Description
赛默飞的乙烯、丙烯中微量CO,CO2分析仪可以在7 min之内完成分析。并且它通过阀切换的配置模式,解决了传统的用一个色谱柱分析乙烯、丙烯中微量CO,CO2分析过程中所带来的分析时间长、镍触媒容易失效以及鬼峰干扰CO定量的问题。此外,通过阀箱安装预分析柱和柱箱安装主分析柱,阀箱、柱箱采用独立控温单元,这种配置,解决了复杂阀系统所带来的老化色谱柱必须拆卸色谱柱,这一问题,降低了维护成本,提高了化验室的工作效率。| Market: | Chemical |
| Keywords: | 气相色谱;CO;CO2;控温阀箱 |
| Matrix: | 乙烯与丙烯 |
For Research Use Only. Not for use in diagnostic procedures.