Find methods for your needs
Assay for Citrate and Phosphate in Pharmaceutical Formulations Using a Compact IC System Be the first to rate this application
Description
Citric acid is a common ingredient found in pharmaceutical formulations. An assay for citrate and phosphate in pharmaceutical formulations using ion chromatography with a Thermo Scientific Dionex IonPac AS11 hydroxide-selective, anion-exchange column and suppressed conductivity detection was validated in Application Note 164. The method incorporates an electrolytic eluent generator to automatically produce an isocratic potassium hydroxide eluent. This record shows that the method can be run using the Thermo Scientific Dionex Integrion system.
Market: Pharma
Keywords: Pharmaceutical, Citric acid, Citrate, Dionex IonPac AS11 column, RFIC, phosphate, Drug, application note 164, Assay, Formulations, Intergrion
Matrix: Drug
Author: Hua Yang
Affiliation: Thermo Fisher Scientific
Uploaded on 1/25/2016.
For Research Use Only. Not for use in diagnostic procedures.