Find methods for your needs
TN44: The Determination of Trace Anions in Concentrated Phosphoric Acid Be the first to rate this application
tn44 determination trace anions concentrated phosphoric acid
Description
The determination of trace anions in phosphoric acid is hampered by a large excess of phosphate ion. Diluting the concentrated sample overcomes the problem of a large concentration of the interfering matrix ion, but lacks the required sensitivity for the contaminant ions of interest. An improved method for determining trace anions in concentrated phosphoric acid has been developed to overcome this problem. This Technical Note describes the theory, set up, and analytical procedure for the determination of chloride, nitrate, and sulfate at sub-mg/L (ppm) levels in 85% (w/w) phosphoric acid.
Market: Chemical
Keywords: Ion Exclusion, IonPac AS11, IonPac ICE-AS6, SEMI, TN 44
Matrix: Phosphoric acid
Author: Edward Kaiser and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 10/2/2015.
For Research Use Only. Not for use in diagnostic procedures.