Find methods for your needs
AN183: Determination of Biogenic Amines in Fermented and Non-Fermented Foods Using Ion Chromatography with Suppressed Conductivity & Integrated Pulsed Amperometric Detections. Be the first to rate this application
an183 determination biogenic amines fermented nonfermented foods using ion chromatography with suppressed conductivity integrated pulsed amperometric detections
Description
The IonPac CS18, a polymeric weak acid cationexchange column, was used to separate biogenic amines in a variety of fermented and non-fermented food samples, with detection by IPAD, suppressed conductivity, and UV. The described method uses a simple electrolytically generated MSA eluent without requiring the use of solvents or aggressive eluent systems that have previously been reported. In addition, the method results in good precision and recovery over a wide range of sample matrices and avoids the need for complex and long derivatization procedures.
Market: Food and Beverage
Keywords: AN 183, Dopamine, Histamine, IonPac CS18, phenylethylamine, Serotonin, Tyramine
Matrix: TCA (trichloroacetic acid) or MSA (methanesulfonic acid)
Uploaded on 6/25/2015.
For Research Use Only. Not for use in diagnostic procedures.