Rate
Ask the Expert
Suggest an Application
Add to Favorites
Create
Parts List
Parts List
Tell a
Friend
Friend
Print
PDF
Find methods for your needs
AN247 (b): Determination of Morpholine, Ethanolamine, and Hydrazine in Simulated Nuclear Power Plant Wastewater
Description
This Application Note describes the determination of μg/L concentrations of hydrazine, morpholine, and ethanolamine (ETA) in a simulated NPP wastewater sample containing mg/L concentrations of common cations. ETA is separated using an IonPac CS15 column with electrolytically generated methanesulfonic acid eluent. See AN 247 (a) for separation of morpholine and hydrazine on an IonPac CS16 column.Market: | Water Analysis |
Keywords: | Amines, ammonium, Magnesium, Nuclear Power Plant, Potassium, Calcium, Lithium, Sodium, ETA, Corrosion Inhibitors, Ethanolamine, IonPac CS15 |
Matrix: | Wastewater |
Author: | Terri Christison, Brian DeBorba, and Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.