Find methods for your needs
AN206: Determination of Oxalate and Other Anions in Bayer Liquor Using Ion Chromatography Be the first to rate this application
an206 determination oxalate other anions bayer liquor using ion chromatography
Description
A direct injection method was designed using a Reagent‑Free™ Ion Chromatography (RFIC™) system with in-line sample cleanup to accurately and reproducibly measure oxalate in Bayer liquor. The online sample preparation removes most of the aluminum so that it does not contaminate either the column or suppressor. Fluoride is used to promote ligand exchange with oxalate for aluminum. This online sample preparation method allows an accurate, reproducible, and rugged determination of oxalate in Bayer liquor.
Market: Material Science
Keywords: Alumina, Aluminium, Aluminum oxalate, Bauxite, Calcination, IonPac AS17, Malonate, Oxalate, RFIC
Matrix: Bayer liquor
Uploaded on 2/2/2015.
For Research Use Only. Not for use in diagnostic procedures.