Rate
Ask the Expert
Suggest an Application
Add to Favorites
Create
Parts List
Parts List
Tell a
Friend
Friend
Print
PDF
Find methods for your needs
AN 165: Determination of Benzoate in Liquid Food Products by Reagent-Free Ion Chromatography
Description
This application describe a simple ion chromatography method for the direct determination of benzoate in liquid food products. Benzoate is separated on a Thermo Scientific Dionex IonPac AS18 column in less than 20 min using a hydroxide gradient followed by suppressed conductivity detection.Market: | Food and Beverage |
Keywords: | AERS 500, benzoate, Diet soda, Flavored soda, Lemon juice, Soy sauce, Thermo Scientific Dionex IonPac AS18 Column |
Matrix: | Liquid food products |
For Research Use Only. Not for use in diagnostic procedures.